Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163233.1 | NP_001156705.1 | 8 - 9 | 1294 | 74 | |
NM_053081.2 | NP_444311.1 | 8 - 9 | 1294 | 74 | ||
GenBank mRNA | AF112439.1 | - | 8 - 9 | 1103 | 74 | |
AY049715.1 | - | 8 - 9 | 1094 | 74 | ||
BC050890.1 | - | 8 - 9 | 1551 | 74 | ||
BC139763.1 | - | 8 - 9 | 1097 | 40110334 | 74 | |
BE292126.1 | - | 3 - 4 | 398 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |