Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027793.1 | NP_082069.1 | 4 - 5 | 298 | 91 | |
GenBank mRNA | AK017352.1 | - | 4 - 5 | 298 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |