Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026236.3 | NP_080512.1 | 14 - 15 | 1493 | 130 | |
GenBank mRNA | AK018396.1 | - | 14 - 15 | 1496 | 130 | |
AK030041.1 | - | 14 - 15 | 1494 | 130 | ||
AK031244.1 | - | 14 - 15 | 1506 | 132 | ||
AK033631.1 | - | 14 - 15 | 1492 | 130 | ||
AK035273.1 | - | 14 - 15 | 1495 | 130 | ||
AK035864.1 | - | 14 - 15 | 1495 | 130 | ||
AK043970.1 | - | 14 - 15 | 1495 | 130 | ||
AK122511.1 | - | 13 - 14 | 2635 | 130 | ||
AK128953.1 | - | 14 - 15 | 1493 | 130 | ||
BC006679.1 | - | 13 - 14 | 1356 | 130 | ||
BC021348.1 | - | 4 - 5 | 1683 | 130 | ||
BC040795.1 | - | 10 - 11 | 1024 | 130 | ||
BC048155.1 | - | 14 - 15 | 1440 | 5698339 | 130 | |
BC050106.1 | - | 14 - 15 | 1511 | 130 | ||
BC062967.1 | - | 14 - 15 | 1497 | 6835444 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |