Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xylb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033209.3 | NP_001028381.1 | 13 - 14 | 1384 | 114 | |
NM_001199568.1 | NP_001186497.1 | 14 - 15 | 1432 | 114 | ||
GenBank mRNA | AK143476.1 | - | 13 - 14 | 1384 | 114 | |
AK164186.1 | - | 14 - 15 | 1226 | 114 | ||
AK165440.1 | - | 13 - 14 | 1181 | 114 | ||
AK165518.1 | - | 13 - 14 | 1181 | 114 | ||
AK165759.1 | - | 14 - 15 | 1228 | 114 | ||
AK167735.1 | - | 4 - 5 | 404 | 114 | ||
AK170197.1 | - | 12 - 13 | 1129 | 114 | ||
BC025442.1 | - | 13 - 14 | 1134 | 114 | ||
BC026406.1 | - | 1 - 2 | 96 | 114 | ||
BC080743.1 | - | 13 - 14 | 1158 | 114 | ||
BC138244.1 | - | 13 - 14 | 1333 | 8861263 | 114 | |
BC138247.1 | - | 13 - 14 | 1333 | 8861266 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |