Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aadat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011834.2 | NP_035964.1 | 8 - 9 | 992 | 76 | |
GenBank mRNA | AF072376.1 | - | 8 - 9 | 992 | 76 | |
AK075578.1 | - | 8 - 9 | 998 | 76 | ||
AK144042.1 | - | 8 - 9 | 945 | 76 | ||
BC012637.1 | - | 8 - 9 | 995 | 4160508 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |