Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eomes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164789.1 | NP_001158261.1 | 2 - 3 | 1513 | 58 | |
NM_010136.3 | NP_034266.2 | 2 - 3 | 1513 | 58 | ||
GenBank mRNA | AB031037.1 | - | 2 - 3 | 1485 | 58 | |
AF013281.1 | - | 2 - 3 | 211 | 58 | ||
AK043327.1 | - | 2 - 3 | 844 | 58 | ||
AK089817.1 | - | 2 - 3 | 1073 | 58 | ||
AK134951.1 | - | 867 | 58 | |||
AK143454.1 | - | 2 - 3 | 834 | 58 | ||
BC094319.1 | - | 2 - 3 | 1358 | 5721456 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Mouse Stem Cell Pluripotency Inventoried Mouse_Stem_Cell Probe spans exons |