Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178633.3 | NP_848748.2 | 813 | 84 | ||
GenBank mRNA | AK032116.1 | - | 361 | 84 | ||
AK167903.1 | - | 604 | 84 | |||
AK220570.1 | - | 813 | 84 | |||
BC031142.1 | - | 684 | 4972965 | 84 | ||
BC031144.1 | - | 707 | 4973325 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |