Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatad2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145596.4 | NP_663571.2 | 7 - 8 | 1293 | 62 | |
GenBank mRNA | AK089074.1 | - | 7 - 8 | 1275 | 62 | |
CA317226.1 | - | 3 - 4 | 274 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |