Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010223.2 | NP_034353.2 | 4 - 5 | 640 | 91 | |
GenBank mRNA | AF030635.1 | - | 3 - 4 | 410 | 91 | |
AY225340.1 | - | 3 - 4 | 523 | 91 | ||
AY278608.1 | - | 4 - 5 | 599 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Detect Genomic DNA Mus musculus Probe spans exons |