Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isyna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023627.1 | NP_076116.1 | 6 - 7 | 829 | 106 | |
GenBank mRNA | AF288525.1 | - | 6 - 7 | 829 | 106 | |
AK005029.2 | - | 6 - 7 | 807 | 106 | ||
AK079323.1 | - | 6 - 7 | 806 | 106 | ||
AK137089.1 | - | 5 - 6 | 911 | 106 | ||
BC003458.1 | - | 6 - 7 | 820 | 2648362 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |