Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028186.4 | NP_082462.3 | 7 - 8 | 800 | 81 | |
GenBank mRNA | AF358136.1 | - | 7 - 8 | 750 | 81 | |
AK008814.1 | - | 7 - 8 | 782 | 81 | ||
AK081904.1 | - | 6 - 7 | 746 | 81 | ||
AK145155.1 | - | 7 - 8 | 782 | 81 | ||
AK146667.1 | - | 7 - 8 | 782 | 81 | ||
AK158350.1 | - | 7 - 8 | 782 | 81 | ||
BC019952.1 | - | 7 - 8 | 769 | 4021772 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |