Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027182.2 | NP_081458.1 | 8 - 9 | 946 | 128 | |
GenBank mRNA | AK010336.1 | - | 8 - 9 | 908 | 128 | |
AK142463.1 | - | 919 | 128 | |||
AK146877.1 | - | 8 - 9 | 947 | 128 | ||
AK151568.1 | - | 8 - 9 | 882 | 128 | ||
BC064315.1 | - | 8 - 9 | 869 | 128 | ||
BC126946.1 | - | 8 - 9 | 845 | 40090292 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |