Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012007.1 | NP_036137.1 | 2 - 3 | 208 | 111 | |
GenBank mRNA | AF136272.1 | - | 2 - 3 | 208 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Mus musculus Probe spans exons |