Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nans | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053179.3 | NP_444409.1 | 3 - 4 | 482 | 135 | |
GenBank mRNA | AB041263.1 | - | 3 - 4 | 522 | 135 | |
AK076290.1 | - | 3 - 4 | 483 | 135 | ||
AK088912.1 | - | 3 - 4 | 492 | 135 | ||
AK156254.1 | - | 3 - 4 | 527 | 135 | ||
AK169211.1 | - | 3 - 4 | 483 | 135 | ||
BC003307.1 | - | 3 - 4 | 575 | 3154822 | 135 | |
BC057977.1 | - | 3 - 4 | 456 | 5052339 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |