Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301412.1 | NP_001288341.1 | 13 - 14 | 1915 | 153 | |
NM_008017.4 | NP_032043.3 | 13 - 14 | 1839 | 153 | ||
GenBank mRNA | AJ534939.1 | - | 12 - 13 | 1670 | 153 | |
AK028233.1 | - | 13 - 14 | 1882 | 153 | ||
AK145341.1 | - | 13 - 14 | 1869 | 153 | ||
AK152090.1 | - | 13 - 14 | 1867 | 153 | ||
BC094380.1 | - | 13 - 14 | 1811 | 30543190 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |