Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138953.2 | NP_620403.2 | 5 - 6 | 909 | 131 | |
GenBank mRNA | AK140906.1 | - | 5 - 6 | 863 | 131 | |
AK144197.1 | - | 5 - 6 | 873 | 131 | ||
AK144978.1 | - | 5 - 6 | 871 | 132 | ||
AK145868.1 | - | 5 - 6 | 858 | 131 | ||
AK154545.1 | - | 5 - 6 | 870 | 131 | ||
AK156164.1 | - | 5 - 6 | 876 | 131 | ||
BC151188.1 | - | 5 - 6 | 864 | 9088077 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |