Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138953.2 | NP_620403.2 | 8 - 9 | 1677 | 67 | |
GenBank mRNA | AK033541.1 | - | 3 - 4 | 665 | 67 | |
AK140906.1 | - | 8 - 9 | 1631 | 67 | ||
AK144197.1 | - | 8 - 9 | 67 | |||
AK144978.1 | - | 8 - 9 | 1641 | 67 | ||
AK145868.1 | - | 8 - 9 | 1626 | 67 | ||
AK154545.1 | - | 8 - 9 | 1638 | 67 | ||
AK156164.1 | - | 8 - 9 | 1644 | 67 | ||
AK161961.1 | - | 1 - 2 | 127 | 67 | ||
BC006925.1 | - | 3 - 4 | 709 | 67 | ||
BC151188.1 | - | 8 - 9 | 1632 | 9088077 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |