Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011078.3 | NP_035208.2 | 16 - 17 | 2300 | 78 | |
GenBank mRNA | AB221590.1 | - | 5 - 6 | 686 | 78 | |
AF043726.1 | - | 16 - 17 | 2300 | 78 | ||
AK172995.1 | - | 14 - 15 | 2007 | 78 | ||
BC051633.1 | - | 16 - 17 | 2298 | 6518591 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |