Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011078.3 | NP_035208.2 | 5 - 6 | 720 | 65 | |
GenBank mRNA | AB221591.1 | - | 5 - 6 | 604 | 65 | |
AF043726.1 | - | 5 - 6 | 720 | 65 | ||
AK172995.1 | - | 3 - 4 | 427 | 65 | ||
BC051633.1 | - | 5 - 6 | 718 | 6518591 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |