Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011078.3 | NP_035208.2 | 8 - 9 | 1161 | 69 | |
GenBank mRNA | AB221591.1 | - | 8 - 9 | 1045 | 69 | |
AF043726.1 | - | 8 - 9 | 1161 | 69 | ||
AK153906.1 | - | 8 - 9 | 1311 | 69 | ||
AK172995.1 | - | 6 - 7 | 868 | 69 | ||
BC051633.1 | - | 8 - 9 | 1159 | 6518591 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |