Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028270.4 | NP_082546.1 | 1 - 2 | 146 | 64 | |
GenBank mRNA | AK088396.1 | - | 1 - 2 | 120 | 64 | |
AK150992.1 | - | 1 - 2 | 48 | 64 | ||
AK151349.1 | - | 1 - 2 | 48 | 64 | ||
AK151364.1 | - | 1 - 2 | 48 | 64 | ||
AK153416.1 | - | 1 - 2 | 48 | 64 | ||
BC020001.1 | - | 1 - 2 | 50 | 4207479 | 64 | |
BC086768.1 | - | 1 - 2 | 105 | 6808412 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |