Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081390.1 | NP_001074859.1 | 9 - 10 | 1871 | 69 | |
NM_001293772.1 | NP_001280701.1 | 9 - 10 | 1875 | 69 | ||
NM_001293773.1 | NP_001280702.1 | 8 - 9 | 1006 | 69 | ||
GenBank mRNA | AF205079.1 | - | 3 - 4 | 299 | 69 | |
AK031696.1 | - | 8 - 9 | 998 | 69 | ||
AK052489.1 | - | 9 - 10 | 1853 | 69 | ||
AK076423.1 | - | 7 - 8 | 643 | 69 | ||
AK087270.1 | - | 8 - 9 | 1008 | 69 | ||
AK173081.1 | - | 8 - 9 | 946 | 69 | ||
BC027364.1 | - | 8 - 9 | 955 | 69 | ||
JX477684.1 | - | 8 - 9 | 998 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |