Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armcx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202500.3 | NP_001189429.2 | 1 - 2 | 115 | 103 | |
GenBank mRNA | CJ167759.1 | - | 1 - 2 | 115 | 103 | |
CJ177091.1 | - | 1 - 2 | 115 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |