Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153585.5 | NP_705813.2 | 16 - 17 | 2139 | 83 | |
GenBank mRNA | AK029103.1 | - | 16 - 17 | 2014 | 83 | |
AK149944.1 | - | 16 - 17 | 1969 | 83 | ||
BC025554.1 | - | 16 - 17 | 2053 | 5321869 | 83 | |
BC036178.1 | - | 13 - 14 | 1595 | 83 | ||
BC039183.1 | - | 16 - 17 | 2113 | 2631793 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |