Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153585.5 | NP_705813.2 | 9 - 10 | 1264 | 66 | |
GenBank mRNA | AK029103.1 | - | 9 - 10 | 1139 | 66 | |
AK149944.1 | - | 9 - 10 | 1097 | 66 | ||
AK157824.1 | - | 9 - 10 | 1163 | 66 | ||
AK161842.1 | - | 9 - 10 | 1186 | 66 | ||
AK171701.1 | - | 9 - 10 | 1244 | 66 | ||
BC025554.1 | - | 9 - 10 | 1125 | 5321869 | 66 | |
BC036178.1 | - | 6 - 7 | 720 | 66 | ||
BC036179.1 | - | 9 - 10 | 1236 | 5059723 | 66 | |
BC039183.1 | - | 9 - 10 | 1238 | 2631793 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |