Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026036.3 | NP_080312.1 | 2 - 3 | 346 | 60 | |
GenBank mRNA | AK012941.1 | - | 2 - 3 | 347 | 60 | |
AK030885.1 | - | 2 - 3 | 326 | 60 | ||
AK128960.1 | - | 2 - 3 | 332 | 60 | ||
AK145743.1 | - | 2 - 3 | 327 | 60 | ||
AK150775.1 | - | 2 - 3 | 347 | 60 | ||
AK151092.1 | - | 2 - 3 | 347 | 60 | ||
AK151431.1 | - | 2 - 3 | 347 | 60 | ||
AK152448.1 | - | 2 - 3 | 348 | 60 | ||
AK152723.1 | - | 2 - 3 | 357 | 60 | ||
AK152817.1 | - | 2 - 3 | 357 | 60 | ||
AK153092.1 | - | 2 - 3 | 347 | 60 | ||
AK154481.1 | - | 2 - 3 | 333 | 60 | ||
AK159439.1 | - | 2 - 3 | 316 | 60 | ||
AK159735.1 | - | 2 - 3 | 341 | 60 | ||
AK165948.1 | - | 2 - 3 | 315 | 60 | ||
AK167499.1 | - | 2 - 3 | 333 | 60 | ||
AK167854.1 | - | 2 - 3 | 338 | 60 | ||
AK170905.1 | - | 2 - 3 | 348 | 60 | ||
AK171394.1 | - | 2 - 3 | 377 | 60 | ||
AY241868.1 | - | 2 - 3 | 383 | 60 | ||
BC027248.1 | - | 2 - 3 | 318 | 2811938 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |