Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Invs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281977.1 | NP_001268906.1 | 9 - 10 | 1423 | 90 | |
NM_001281978.1 | NP_001268907.1 | 11 - 12 | 1701 | 90 | ||
NM_010569.4 | NP_034699.3 | 9 - 10 | 1475 | 90 | ||
GenBank mRNA | AF034860.3 | - | 9 - 10 | 1417 | 90 | |
AJ010902.1 | - | 9 - 10 | 1416 | 90 | ||
AK041671.1 | - | 8 - 9 | 1242 | 90 | ||
AK158766.1 | - | 11 - 12 | 1660 | 90 | ||
BC126929.1 | - | 9 - 10 | 1312 | 40081431 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |