Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Umodl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177465.4 | NP_803416.2 | 14 - 15 | 2856 | 67 | |
GenBank mRNA | AY771616.1 | - | 15 - 16 | 2773 | 67 | |
AY771617.1 | - | 14 - 15 | 2860 | 67 | ||
AY771618.1 | - | 14 - 15 | 2515 | 67 | ||
BC051427.1 | - | 14 - 15 | 2511 | 67 | ||
BC132381.1 | - | 1 - 2 | 45 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |