Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Umodl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177465.4 | NP_803416.2 | 16 - 17 | 3310 | 101 | |
GenBank mRNA | AK040446.1 | - | 8 - 9 | 1811 | 101 | |
AY771616.1 | - | 17 - 18 | 3227 | 101 | ||
AY771617.1 | - | 16 - 17 | 3314 | 101 | ||
AY771618.1 | - | 16 - 17 | 2969 | 101 | ||
BC051427.1 | - | 16 - 17 | 2965 | 101 | ||
BC132381.1 | - | 3 - 4 | 499 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |