Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009654.3 | NP_033784.2 | 6 - 7 | 754 | 92 | |
GenBank mRNA | AJ011413.1 | - | 6 - 7 | 741 | 92 | |
AJ457860.1 | - | 6 - 7 | 708 | 92 | ||
AK010025.1 | - | 6 - 7 | 743 | 92 | ||
AK050248.1 | - | 6 - 7 | 743 | 92 | ||
AK050644.1 | - | 5 - 6 | 612 | 92 | ||
AK131891.1 | - | 6 - 7 | 746 | 92 | ||
AK160487.1 | - | 6 - 7 | 746 | 92 | ||
BC024643.1 | - | 6 - 7 | 728 | 4242076 | 92 | |
BC049971.1 | - | 6 - 7 | 728 | 4161986 | 92 | |
M16111.1 | - | 3 - 4 | 414 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |