Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Secisbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK030021.1 | - | 13 - 14 | 1927 | 71 | |
AK156076.1 | - | 13 - 14 | 2003 | 71 | ||
AK168634.1 | - | 13 - 14 | 1960 | 71 | ||
BC038877.1 | - | 1 - 2 | 73 | 71 | ||
BC125391.1 | - | 13 - 14 | 1983 | 40129906 | 71 | |
BC125393.1 | - | 13 - 14 | 1980 | 40129908 | 71 | |
BC144795.1 | - | 13 - 14 | 1978 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |