Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400014745.mm.1 |
NM_001081174.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014745.mm.1 |
NM_001081174.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014745.mm.1 |
TC0400001782.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014753.mm.1 |
TC0400001782.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014746.mm.1 |
NM_001081174.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014746.mm.1 |
NM_001081174.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014746.mm.1 |
TC0400001782.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014744.mm.1 |
NM_001081174.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014744.mm.1 |
NM_001081174.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014744.mm.1 |
TC0400001782.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014757.mm.1 |
TC0400001782.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014747.mm.1 |
NM_001081174.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014747.mm.1 |
NM_001081174.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014747.mm.1 |
TC0400001782.mm.1 |
MTA TC |