Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400014505.mm.1 |
NM_031873.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014505.mm.1 |
NM_031873.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014505.mm.1 |
TC0400004197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014507.mm.1 |
NM_031873.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014507.mm.1 |
NM_031873.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014507.mm.1 |
TC0400004197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014506.mm.1 |
NM_031873.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014506.mm.1 |
NM_031873.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014506.mm.1 |
TC0400004197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014508.mm.1 |
NM_031873.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014508.mm.1 |
NM_031873.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014508.mm.1 |
TC0400004197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014502.mm.1 |
NM_031873.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014502.mm.1 |
NM_031873.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014502.mm.1 |
TC0400004197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014504.mm.1 |
NM_031873.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014504.mm.1 |
NM_031873.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014504.mm.1 |
TC0400004197.mm.1 |
MTA TC |