Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr7a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025337.3 | NP_079613.3 | 6 - 7 | 72 | ||
GenBank mRNA | AF525358.2 | - | 6 - 7 | 72 | ||
AJ271800.1 | - | 6 - 7 | 72 | |||
AJ271801.1 | - | 6 - 7 | 72 | |||
AK002664.1 | - | 6 - 7 | 72 | |||
AK003915.1 | - | 6 - 7 | 72 | |||
AK143203.1 | - | 6 - 7 | 72 | |||
BC031857.1 | - | 6 - 7 | 5124841 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |