Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167994.1 | NP_001161466.1 | 10 - 11 | 1262 | 94 | |
NM_172540.2 | NP_766128.2 | 9 - 10 | 1186 | 94 | ||
GenBank mRNA | AK028999.1 | - | 9 - 10 | 1142 | 94 | |
AK049523.1 | - | 9 - 10 | 1184 | 94 | ||
AK137595.1 | - | 1148 | 94 | |||
BC030387.1 | - | 8 - 9 | 847 | 94 | ||
BC056937.1 | - | 10 - 11 | 1203 | 5716144 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |