Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010139.3 | NP_034269.2 | 1 - 2 | 216 | 113 | |
GenBank mRNA | AK137704.1 | - | 1 - 2 | 198 | 113 | |
AK144202.1 | - | 1 - 2 | 200 | 113 | ||
BC140960.1 | - | 1 - 2 | 172 | 9055572 | 113 | |
BY024029.1 | - | 1 - 2 | 197 | 113 | ||
CJ088701.1 | - | 1 - 2 | 220 | 113 | ||
U07634.1 | - | 1 - 2 | 180 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |