Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010139.3 | NP_034269.2 | 6 - 7 | 1569 | 65 | |
GenBank mRNA | AK137704.1 | - | 6 - 7 | 1551 | 65 | |
AK144202.1 | - | 6 - 7 | 1553 | 65 | ||
BC140960.1 | - | 6 - 7 | 1525 | 9055572 | 65 | |
X78339.1 | - | 6 - 7 | 1531 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |