Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033263.5 | NP_001028435.1 | 12 - 13 | 3128 | 117 | |
NM_001301014.1 | NP_001287943.1 | 12 - 13 | 3128 | 117 | ||
GenBank mRNA | AK147446.1 | - | 12 - 13 | 3129 | 117 | |
AK220325.1 | - | 12 - 13 | 2003 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |