Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC058120.1 | - | 1 - 2 | 125 | 6810624 | 95 |
BC138330.1 | - | 1 - 2 | 103 | 8861350 | 95 | |
BC138331.1 | - | 1 - 2 | 103 | 8861351 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |