Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021604.3 | NP_067617.3 | 11 - 12 | 2224 | 108 | |
GenBank mRNA | AK147577.1 | - | 10 - 11 | 2245 | 108 | |
BC058651.1 | - | 10 - 11 | 2337 | 108 | ||
BC150703.1 | - | 9 - 10 | 1678 | 9087614 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |