Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK039305.1 | - | 1 - 2 | 100 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |