Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013901.2 | NP_038929.2 | 2 - 3 | 279 | 62 | |
GenBank mRNA | AK146175.1 | - | 2 - 3 | 271 | 62 | |
AK159290.1 | - | 2 - 3 | 136 | 62 | ||
BC005474.1 | - | 2 - 3 | 241 | 3591856 | 62 | |
BC132461.1 | - | 1 - 2 | 216 | 40130738 | 62 | |
BC138459.1 | - | 1 - 2 | 216 | 8861479 | 62 | |
BC147738.1 | - | 1 - 2 | 217 | 9056687 | 62 | |
BC147744.1 | - | 1 - 2 | 217 | 9056693 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |