Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00000716.mm.1 |
NM_199063.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000716.mm.1 |
NM_199063.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000716.mm.1 |
TC0X00003386.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000719.mm.1 |
NM_199063.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000719.mm.1 |
NM_199063.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000719.mm.1 |
TC0X00003386.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000715.mm.1 |
NM_199063.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000715.mm.1 |
NM_199063.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000715.mm.1 |
TC0X00003386.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000717.mm.1 |
NM_199063.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000717.mm.1 |
NM_199063.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000717.mm.1 |
TC0X00003386.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000718.mm.1 |
NM_199063.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000718.mm.1 |
NM_199063.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000718.mm.1 |
TC0X00003386.mm.1 |
MTA TC |