Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039186.1 | NP_001034275.1 | 6 - 7 | 1448 | 111 | |
NM_001039187.1 | NP_001034276.1 | 4 - 5 | 908 | 111 | ||
GenBank mRNA | AB236329.1 | - | 4 - 5 | 817 | 111 | |
BC114998.1 | - | 2 - 3 | 380 | 111 | ||
BC121189.1 | - | 5 - 6 | 1005 | 111 | ||
M77196.1 | - | 6 - 7 | 1426 | 111 | ||
X15351.1 | - | 6 - 7 | 1460 | 111 | ||
X53084.1 | - | 4 - 5 | 837 | 111 | ||
X67278.1 | - | 4 - 5 | 920 | 111 | ||
X67280.1 | - | 6 - 7 | 1377 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |