Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec61g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109971.1 | NP_001103441.1 | 2 - 3 | 504 | 149 | |
GenBank mRNA | AK012393.1 | - | 2 - 3 | 122 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |