Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198027.2 | NP_932144.2 | 5 - 6 | 439 | 77 | |
GenBank mRNA | AK209174.1 | - | 1 - 2 | 87 | 77 | |
BC029805.1 | - | 5 - 6 | 391 | 4952089 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |