Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nanog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289830.1 | NP_001276759.1 | 4 - 5 | 525 | 105 | |
NM_001289831.1 | NP_001276760.1 | 4 - 5 | 509 | 105 | ||
NM_028016.3 | NP_082292.1 | 3 - 4 | 728 | 105 | ||
GenBank mRNA | AB093574.1 | - | 3 - 4 | 699 | 105 | |
AB126939.1 | - | 3 - 4 | 510 | 105 | ||
AF507043.1 | - | 3 - 4 | 700 | 105 | ||
AK010332.1 | - | 3 - 4 | 700 | 105 | ||
AK167259.1 | - | 3 - 4 | 699 | 106 | ||
AY278951.1 | - | 3 - 4 | 577 | 105 | ||
AY455282.1 | - | 4 - 5 | 509 | 105 | ||
AY455285.1 | - | 4 - 5 | 525 | 105 | ||
BC132017.1 | - | 3 - 4 | 725 | 40130294 | 105 | |
BC137873.1 | - | 3 - 4 | 725 | 8860892 | 105 | |
BC144996.1 | - | 3 - 4 | 725 | 9053544 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |