Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | UBX domain protein 7 | Ubxn7 |
A630090P18, AI196514, Ubxd7, mKIAA0794 |
Sense | Chr. 16 - 32332184 - 32393750 on NCBI Build GRCm38 | |
Downstream | transmembrane 4 L six family member 19 | Tm4sf19 | EG277203 | 37.7kb | Sense | Chr. 16 - 32400012 - 32408579 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |