Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173399.3 | NP_775575.2 | 1 - 2 | 191 | 84 | |
GenBank mRNA | AK031172.1 | - | 1 - 2 | 122 | 85 | |
BC054551.1 | - | 1 - 2 | 49 | 6400036 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |