Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phyhipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178621.4 | NP_848736.2 | 1 - 2 | 291 | 89 | |
GenBank mRNA | AK029522.1 | - | 1 - 2 | 278 | 89 | |
AK032166.1 | - | 1 - 2 | 292 | 89 | ||
AK034448.1 | - | 1 - 2 | 272 | 89 | ||
AK083011.1 | - | 1 - 2 | 292 | 89 | ||
AK164118.1 | - | 1 - 2 | 291 | 89 | ||
BC037618.1 | - | 1 - 2 | 279 | 5400532 | 89 | |
BC124999.1 | - | 1 - 2 | 89 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |